missing translation for 'onlineSavingsMsg'
Learn More

Orotic acid, anhydrous, 97%

Catalog No. AAB2534936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2534936 500 g
AAB2534922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2534936 Supplier Thermo Scientific Chemicals Supplier No. B2534936

missing translation for 'validMainframeMsg'

CAS: 65-86-1 | C5H4N2O4 | 156.097 g/mol

Orotic acid is an intermediate in de novo pyrimidine biosynthesis that may be used to study the specificity and kinetics of orotate phosphoribosyltransferase (OPRT) which catalyzes the reversible phosphoribosyl transfer from 5?-phospho-α-d-ribose 1?-diphosphate (PRPP) to orotic acid (OA), forming pyrophosphate and orotidine 5?-monophosphate (OMP). It is used as a starting material for the potential commercial bioproduction of uridine 5?-monophosphate (UMP) by microbes such as Corynebacterium ammoniagenes (ATCC 6872) or Saccharomyces cerevisiae. It may be used to study the AMPK/SREBP-1 dependent cell signaling pathway and transcription regulation mechanisms that induce hepatic lipogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Orotic acid is an intermediate in de novo pyrimidine biosynthesis that may be used to study the specificity and kinetics of orotate phosphoribosyltransferase (OPRT) which catalyzes the reversible phosphoribosyl transfer from 5′-phospho-α-d-ribose 1′-diphosphate (PRPP) to orotic acid (OA), forming pyrophosphate and orotidine 5′-monophosphate (OMP). It is used as a starting material for the potential commercial bioproduction of uridine 5′-monophosphate (UMP) by microbes such as Corynebacterium ammoniagenes (ATCC 6872) or Saccharomyces cerevisiae. It may be used to study the AMPK/SREBP-1 dependent cell signaling pathway and transcription regulation mechanisms that induce hepatic lipogenesis.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 65-86-1
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.097
MDL Number MFCD00006027
InChI Key PXQPEWDEAKTCGB-UHFFFAOYSA-N
Synonym orotic acid, 6-carboxyuracil, orodin, oropur, orotonin, orotonsan, oroturic, orotyl, vitamin b13, whey factor
PubChem CID 967
ChEBI CHEBI:16742
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid
SMILES C1=C(NC(=O)NC1=O)C(=O)O

Specifications

Melting Point 344°C to 347°C
Quantity 500 g
Solubility Information Slightly soluble in water.
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material Orotic acid, anhydrous
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-619-8
RTECSNumber RM3180000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only