missing translation for 'onlineSavingsMsg'
Learn More

Ornidazole, 99%

Catalog No. p-7042601
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6310006 5 g
AAJ6310014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6310006 Supplier Thermo Scientific Chemicals Supplier No. J6310006
Only null left

Interferes with electron transport in metabolic pathways | CAS: 16773-42-5 | C7H10ClN3O3 | 219.625 g/mol

Ornidazole is a drug that cures some protozoan infections. It has been investigated for use in Crohn's disease after bowel resection. It is indicated in treatment of mixed amoebiasis, mixed amoebic dysentery, mixed giardiasis, trichomoniasis, bacterial vaginosis, Sexually transmitted diseases, infections of gynaecology, lower respiratory tract infections, ENT, surgical and dental infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 16773-42-5
Molecular Formula C7H10ClN3O3
Molecular Weight (g/mol) 219.625
MDL Number MFCD00057960
InChI Key IPWKIXLWTCNBKN-UHFFFAOYSA-N
Synonym ornidazole, madelen, tiberal, ornidazolum, 1-3-chloro-2-hydroxypropyl-2-methyl-5-nitroimidazole, ornidazol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl propan-2-ol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl-2-propanol, ornidal, 1-2-hydroxy-3-chloropropyl-2-methyl-5-nitroimidazole
PubChem CID 28061
ChEBI CHEBI:75176
IUPAC Name 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
SMILES CC1=NC=C(N1CC(CCl)O)[N+](=O)[O-]

Specifications

Melting Point 84°C to 86°C
Color White to Yellow
Quantity 5 g
Merck Index 14,6872
Solubility Information Soluble in ethanol at 50mg/ml
Formula Weight 219.63
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ornidazole
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 240-826-0
RTECSNumber NI5460000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only