Learn More
Perkin Elmer Canada Flared PVC Two-Stop Peristaltic Pump Tubing - 0.38 mm I.D., Orange/Green, Pkg. 12 DFS Item
Supplier:  Perkin Elmer Canada N0777042
This flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-905-2406
Only null left
Explore available promotions