Learn More
Thermo Scientific™ Orion™ Universal Power Adapter for Orion™ Versa Star Meters
Description
Ensure a constant power supply to your meter using a spare or replacement Thermo Scientific™ Orion™ Universal Power Adapter for the Thermo Scientific™ Orion™ Versa Star meters.
Specifications
Specifications
| Description | Orion Versa Star universal power adapter |
| Type | Meter Accessories |
| Electrical Requirements | 100/240 VAC, 50/60 Hz |
| Frequency | 50/60 Hz |
| Includes | Universal power adapter with US, EU, UK and China plug plates |
| Voltage | 100/240 VAC |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.