missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion Star™ A221 Portable pH Meter

Catalog No. 13303212
Change view
Click to view available options
Type:
pH Meter Food/Beverage Kit
pH Meter ORP Kit
pH Meter Standard Kit
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13303212 pH Meter Food/Beverage Kit
13645522 -
13303210 pH Meter Standard Kit
13303211 pH Meter ORP Kit
13645524 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13303212 Supplier Thermo Scientific™ Supplier No. STARA2218

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reliably measure pH, mV, ORP and temperature with our portable pH meter for advanced field analysis and printer or computer interfacing.

Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A221 Portable pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the field. Perform up to a 5 point pH calibration, log up to 1000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Waterproof meter ensures durable, rugged performance in outdoor environments with an IP67-rated housing.

Don't leave your measurements to chance when analyzing samples outside the lab. Orion Star A221 Portable pH Meters are designed to offer the reliability you need in a user-friendly package for your dedicated pH testing needs. This waterproof, portable meter displays critical data with your measurement including temperature, date and time, read type, measurement stability, active calibration info and power source status.

  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading and Continuous shows changing readings with hold option
  • Data log collects and date/time stamps up to 1000 measurement sets
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty

Specifications

Description Orion Star A221 pH portable meter food kit with ROSS Spear-Tip pH electrode and ATC probe
Accuracy (mV) ±0.2 or ±0.05%, whichever is greater
Data Management Details Transfer single, range or all data points to printer or computer
Accuracy (Temperature) ±0.1°C
Accuracy (pH) ±0.002
Outputs USB, RS-232
Includes Star A221 pH meter, 8163BNWP ROSS spear-tip pH electrode, 927007MD stainless steel ATC probe, pH 4/7/10 buffer and rinse pouches, ROSS storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable
Measurement Modes pH, mV, Relative mV, ORP with Temperature
Calibration pH, relative mV (RmV), ORP and temperature
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) ±2000
Range (ORP) ±2000.0mV
Resolution (mV) 0.1mV
Calibration Points 1 to 5
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 22.0°F to 221.0°F
Resolution (Temperature) 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Temperature Probe Calibration 1 point temperature offset calibration
Display Type LCD, Graphic with Backlight
Backlight Option Yes (Selectable)
Memory Non-volatile memory preserves data log, calibration log and meter settings
Data Points 1000 with Date and Time Stamp
Log Function Type Automatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
Alarm Output Calibration due alarm (fixed 12 hour)
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP67
AC Adapter Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Dimensions (L x W x H) 24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
Weight (Metric) 0.5 kg
Weight (English) 1.2 lb.
Keypad Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Model Orion Star A221 pH/mV/RmV/ORP/Temperature Portable Meter
Power Supply 4 AA batteries (included) or universal AC adapter (sold separately)
Probe Type pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (includes 8163BNWP ROSS spear-tip pH electrode and 927007MD ATC probe)
Languages English, Spanish, French, Italian, German, Chinese, Portuguese and Korean available with software update
Range (Relative mV) ±2000.0mV
Relative Humidity Range 5 to 85%, non-condensing
Isopotential Point 7.000pH
Stability Smart Stability and Smart Averaging
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Channels 1
Battery Type 4 x AA
Type pH Meter Food/Beverage Kit
Battery Life 800 hr.
Show More Show Less
How long is the warranty period for Orion pH meters?

At Thermo Fisher Scientific, we stand behind the quality and reliability of our instruments. That's why we offer a minimum two-year (2) warranty on all Orion benchtop and portable pH meters. Obtain peace of mind knowing that your investment is protected. Should any issues arise within the warranty period, our team of experts is committed to providing you with prompt and efficient service. Be sure to check instrument specifications to confirm the warranty coverage period.

How do I perform a calibration using Orion pH meters?

Calibration is an important step to obtaining accurate results on your Orion benchtop or portable pH meter. Before beginning the calibration process, ensure that you are utilizing freshly poured, non-expired calibration buffers to help ensure accurate results. Follow the steps below to calibrate your pH meter.

• Prepare the calibration solutions: Obtain at least two pH reference solutions with known pH values. Ideally, use one solution close to the expected pH of your sample, and another solution at a different pH value. Ensure that the solutions are fresh and have not expired.
• Clean the electrode: Rinse the electrode with distilled or deionized water to remove any debris or contaminants. If necessary, gently wipe the electrode with a soft cloth or tissue.
• Turn on the pH meter: Power on the pH meter and allow it to warm up, if needed.
• Immerse the electrode: Immerse the electrode into a beaker containing the first calibration solution. Ensure that the electrode is fully submerged and that it is not touching the sides or bottom of the beaker.
• Allow stabilization: Allow the pH reading to stabilize. This may take a few moments, depending on the specific meter and solution being used. The reading should become steady and remain constant.
• Adjust the pH value: Using the calibration controls on the pH meter, adjust the reading to match the known pH value of the calibration solution. Reference the user manual for specific guidance on adjusting the pH.
• Rinse the electrode: After calibration, rinse the electrode with distilled or deionized water to remove any residual calibration solution.
• Repeat calibration: Repeat the calibration steps with the second calibration solution, following the same process as before.
• Verify accuracy: After calibration, it is recommended to check the pH meter's accuracy by measuring a pH solution of known value. This can help ensure the reliability of your measurements.

To learn more about pH meter calibration, download our technical note: Understanding Calibration of Laboratory pH Systems. See a live calibration by watching our video: Understanding Calibration of Laboratory pH Systems.

How do I perform an analysis using Orion pH meters?

Before beginning an analysis on your Orion benchtop or portable pH meter, assemble your materials. You will need a pH electrode suitable for your application, calibration buffer solutions, and a test sample. Follow the steps below to obtain your pH analysis.

• Calibrate the pH meter: Start by calibrating your pH meter using the calibration buffer solutions. Typically, you will need pH 4, pH 7, and pH 10. Be sure to reference the user manual for instructions on how to properly calibrate your pH meter.
• Prepare your sample: Ensure that your sample is at the desired temperature (most frequently room temperature). If necessary, filter the sample to remove any particles or impurities that could impact the pH reading.
• Immerse the electrode: Dip the pH electrode into the sample. Avoid touching the sides or bottom of the container to prevent inaccurate readings.
• Allow for stabilization: Give the pH meter some time to stabilize and reach a steady reading. This may take a few seconds to a minute depending on your meter and the characteristics of the sample.
• Record the pH value: Once the pH meter stabilizes, record the pH value displayed on the meter. If applicable, also record the temperature value.
• Rinse the electrode: After each measurement, rinse the electrode with distilled or deionized water to remove any residue or contaminants. This helps ensure accurate subsequent readings.
• Repeat (if necessary): If you need to analyze multiple samples, repeat the process with each sample, recalibrating the pH meter if required.

To learn more about how to perform a pH analysis, watch our video:Measuring Drinking Water Using the Orion Versa Star Pro Bench Meter

How do I choose an Orion pH electrode for my application?

When choosing an Orion pH electrode for your application there are many factors to consider. Sample type, container dimensions, accuracy/ precision requirements, junction design, durability, maintenance, and meter compatibility are all key features to evaluate.
To learn more about how to choose the right electrode for your application, watch our videos: How to Choose the Right pH Electrode and Understanding Your Sample to Select the Right pH Electrode

What supplies do I need to perform an analysis with Orion pH meters?

To perform an analysis using Orion benchtop and portable pH meters, you will need the following supplies: pH electrode (or micro electrode for small sample volumes/ containers), calibration buffer solutions, distilled or deionized water, sample containers, stirrer probe (optional), and electrode cleaning solution(s) (s). For your convenience, our all-in-one pH buffer kits can be purchased, which include pH buffers (4.01 pH, 7.00 pH, and 10.01 pH), electrode cleaning solution, and electrode and storage solution.

How are Orion pH meters powered?

Orion benchtop and portable pH meters are powered differently to meet the needs of your testing environment. Orion portable pH meters are powered by four (4) AA batteries (included), or a universal AC adapter (sold separately). Orion benchtop pH meters are powered by an AC adapter typically consisting of a universal 100-240 V 50/60 Hz power supply and include interchangeable wall plugs for US/Japan, Europe, UK/Singapore, Australia/New Zealand and China outlets.

Are Orion pH meters splash and dustproof?

Orion benchtop and portable pH meters are designed to be splash and dustproof. They are built with durable materials containing protective features to help ensure reliable performance. Our portable meters are IP-67 rated to protect against splashes, dust and limited immersion. Our benchtop meters are IP-54 rated to protect against splashes and dust. Please note that while our pH meters are resistant to splashes and dust, they are not fully waterproof. Reference the user manual for additional details regarding proper usage and maintenance.

What additional measurement parameters are available on Orion pH meters?

Orion benchtop and portable multiparameter meters offer the ability to measure additional parameters beyond pH. Dependent upon the meter model selected, users can also monitor conductivity, dissolved oxygen (DO), ion concentration and more.
To shop the full range of Orion electrochemistry meters, click here

How many measurement channels are available on Orion pH meters?

Orion benchtop and portable pH meters come with a variety of measurement channel options designed to support your needs. Choose from as little as one (1) measurement channel for more streamlined applications, to as many as four (4) measurement channels for more complex workflows where multi-parameter or concurrent analysis is a requirement.
To shop Orion multiparameter electrochemistry meters, click here

How accurate are Orion pH meters?

The pH accuracy on Orion benchtop and portable pH meters varies upon the meter model selected. Generally, Orion pH meters offer a range of ±0.002 to 0.01. Be sure to check instrument specifications to ensure your desired pH accuracy can be achieved.
To learn more about how to get accurate pH measurements, watch our video: 7 Common Mistakes When Measuring pH

What is the pH range on Orion pH meters?

The pH range on Orion benchtop and portable pH meters varies upon the meter model selected. Generally, Orion pH meters offer a range of -2.000 to 20.000 pH. Be sure to check instrument specifications to ensure your desired pH range is covered.
To learn more about how to get accurate pH measurements, watch our video:7 Common Mistakes When Measuring pH

How do I choose between an Orion benchtop or portable pH meter?

When choosing between an Orion benchtop or portable pH meter, consider factors such as portability needs, measurement capabilities, accuracy requirements, desired features, and budget constraints. Our portable meters offer convenience for field use, while our benchtop meters provide higher accuracy and contain a more robust feature offering. Consider your specific application needs before selecting a pH meter.
To learn more on how to choose between a benchtop or portable pH meter, watch our videos: How to Choose a Benchtop pH Meter and How to Choose a Portable pH Meter

What applications can benefit from pH analysis?

pH analysis is beneficial to a wide range of applications across various industries and scientific fields. Some key applications that can benefit from pH analysis include environmental monitoring, municipal water treatment, agriculture, food and beverage, industrial processes, pharmaceutical, healthcare and scientific research.
Browse our Water Quality Resource Library to learn more via application notes, technical notes, videos and more.

How does a pH meter work?

A pH meter uses electrochemistry to measure the acidity or alkalinity of a sample. It consists of a glass electrode and a reference electrode that measure the sample and perform the analysis. The glass electrode is sensitive to hydrogen ions (H+) in a sample. When the glass electrode is immersed in the sample, it generates a small electrical potential based on the concentration of hydrogen ions. This potential is then measured and compared to the potential of the reference electrode, which provides a stable reference point. The difference in potentials between the two electrodes is then used to calculate the pH and displayed as a reading on the meter.
To learn more about pH analysis, download our eBook: pH Measurement Handbook.