missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Silver/Sulphide Ion Selective Electrodes

Catalog No. 13642266
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13642266 BNC (Waterproof)
13641949 BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13642266 Supplier Thermo Scientific™ Supplier No. 9616BNWP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Achieve fast, precise, and cost-effective silver and sulfide ion measurements with dual-electrode technology. Perfect for low-level cyanide and halide titrations.

Make quick, simple, accurate and economical measurements of silver and sulfide ions in aqueous solutions with Thermo Scientific™ Orion™ Silver/Sulfide Electrodes. These electrodes have dual electrode-measuring silver and sulfide ions in aqueous solutions, and perform low-level cyanide and halide titrations.

The ThermoFisher Orion Silver/Sulfide electrode complies with SM 4500-S2- G, ASTM D4658-08 for Wastewater measurements.

Features:

  • 9616BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
  • Silver/Sulfide Half-Cell Electrode 9416BN is to be used with the Orion 900200 sure flow double Junction reference electrode.

Solutions required

  • ISA
    • 940011 ISA for silver measurements, 475 mL bottles
    • 941609 SAOB II reagent pack for sulfide measurements, 4 x 475 mL bottles
  • Electrode fill solutions 5 x 60 mL bottles
    • 900062 Optimum Results B for most silver or sulfide measurements and titrations
    • 900061 Optimum Results A for precise sulfide measurements and titrations
    • 900067 Optimum Results C for precise silver measurements and titrations
  • Electrode fill solution for double junction reference half cell electrode
    • 900002 Inner chamber filling solution, 5 x 60 mL bottles
    • 900003 Outer chamber filling solution, 5 x 60 mL bottles
  • Titrant for sulphide titration
    • 948206 0.1 M lead perchlorate solution, 475 mL bottles

Items included with

9619BNWP

948201: Polishing strip. Qty 2.

900062: Optimum Results B, 60 mL 1 bottle

9419BN

948201: Polishing strip. Qty 2.

Warranty period

12 months from date of purchase

Specifications

Applications Potentiometric titrations for chloride and other halides; silver by titration or direct ISE reading.
Connector Types BNC (Waterproof)
Description Ionplus Sure-Flow solid state combination with WPBNC
Drift Drift-free Potentials
Junction Sure-Flow Junction
Length (English) Cable 3 ft.
Length (Metric) Cable 1 m
Material Epoxy
Certifications/Compliance USP 541, SM 4500-Cl D, AOAC 935.47, AOAC 937.09, AOAC 983.14, EN12133, ISO 5943
For Use With (Equipment) Orion Star Series Meters
Temperature (Metric) Operating 0°C to 80°C
Type Combination ISE
Show More Show Less