missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Silver/Sulphide Ion Selective Electrodes

Catalog No. 13641949
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13641949 BNC
13642266 BNC (Waterproof)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13641949 Supplier Thermo Scientific™ Supplier No. 9416BN

Achieve fast, precise, and cost-effective silver and sulfide ion measurements with dual-electrode technology. Perfect for low-level cyanide and halide titrations.

Make quick, simple, accurate and economical measurements of silver and sulfide ions in aqueous solutions with Thermo Scientific™ Orion™ Silver/Sulfide Electrodes. These electrodes have dual electrode-measuring silver and sulfide ions in aqueous solutions, and perform low-level cyanide and halide titrations.

The ThermoFisher Orion Silver/Sulfide electrode complies with SM 4500-S2- G, ASTM D4658-08 for Wastewater measurements.

Features:

  • 9616BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
  • Silver/Sulfide Half-Cell Electrode 9416BN is to be used with the Orion 900200 sure flow double Junction reference electrode.

Solutions required

  • ISA
    • 940011 ISA for silver measurements, 475 mL bottles
    • 941609 SAOB II reagent pack for sulfide measurements, 4 x 475 mL bottles
  • Electrode fill solutions 5 x 60 mL bottles
    • 900062 Optimum Results B for most silver or sulfide measurements and titrations
    • 900061 Optimum Results A for precise sulfide measurements and titrations
    • 900067 Optimum Results C for precise silver measurements and titrations
  • Electrode fill solution for double junction reference half cell electrode
    • 900002 Inner chamber filling solution, 5 x 60 mL bottles
    • 900003 Outer chamber filling solution, 5 x 60 mL bottles
  • Titrant for sulphide titration
    • 948206 0.1 M lead perchlorate solution, 475 mL bottles

Items included with

9619BNWP

948201: Polishing strip. Qty 2.

900062: Optimum Results B, 60 mL 1 bottle

9419BN

948201: Polishing strip. Qty 2.

Warranty period

12 months from date of purchase

Specifications

Connector Types BNC
Description Solid state half-cell with BNC connector
Drift Drift-free Potentials
Junction Sure-Flow Junction
Length (English) Cable 3 ft.
Length (Metric) Cable 1 m
For Use With (Equipment) ISE Meter
Temperature (Metric) Operating 0°C to 80°C
Type Half-cell Ion Selective Electrode