missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Epoxy Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.

Supplier:  Thermo Scientific™ 8107BNURCA

 View more versions of this product

Catalog No. 13642675


Only null left

Description

Description

Thermo Scientific™ Orion™ ROSS Ultra Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra line with a built-in ATC. These products offer a low-maintenance, gel-filled, epoxy body and are best used where minimal maintenance is required. Non-refillable Orion ROSS Ultra Triodes come with an 18 month warranty.
 

Measure pH and Temperature with One Low-Maintenance Electrode

Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance. Sluggish response, poor accuracy and drift are common. The Thermo Scientific Orion ROSS Ultra 8107 Triode pH electrode responds within one minute, proving a stable, accurate reading.

Complete your pH measurements faster. Get long, maintenance-free electrode lifetime with special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.

Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.

Target Markets:

  • Pharmaceutical/Biopharmaceutical
  • Food Manufacturing
  • Academic/Government
  • Environmental
Applications:
  • Quality Control
  • Research
  • General Purpose
Specifications

Chemical Identifiers

Glass Capillary
0 to 14
pH/ATC Combination Electrode
ROSS
0.03 pH for buffers using automatic temperature compensation
18 Months
12 mm
Rugged Epoxy Body, Filled with Low Maintenance Gel
0°C to 80°C
None required
7
0.01
120 mm

Specifications

Orion ROSS Ultra Low Maintenance pH/ATC Triode Combination Electrodes with a BNC and RCA Connector and 1 meter cable
Glass Capillary
0 to 14
pH/ATC Combination Electrode
Mettler Toledo Five and Seven Series pH Meters
7
0.01
120 mm
12 mm
BNC (Waterproof), RCA
Rugged Epoxy Body, Filled with Low Maintenance Gel
0°C to 80°C
None required
ROSS
0.03 pH for buffers using automatic temperature compensation
18 Months
1 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions