Learn More
Thermo Scientific™ Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Catalog No. 0935744
Change view
Click to view available options
Type:
Calibration Resistor Kit
Replacement Flow Cell
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
0935744 Replacement Flow Cell
13303154 Calibration Resistor Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0935744 Supplier Thermo Scientific™ Supplier No. 013017
Perform virtually any electrochemistry analysis with these conductivity standards, solutions and accessories.
Confidently perform conductivity tests and measurements with Thermo Scientific™ Orion™ Accessories. Find an additional or replacement Glass Flow Cell for use with the Orion 2-Cell Ultra-pure Conductivity Probe. Use the Thermo Scientific Orion Conductivity Resistor Calibration Kit to verify the performance of Thermo Scientific™ Orion™ Versa Star Pro™ and Orion Star™ Bench and Portable Conductivity Meters. Each conductivity calibration kit includes six resistors that simulate nominal conductance values between 1 μS and 100 mS and have 8 pin MiniDIN connectors.
Conductivity Standards
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
Specifications
Description Replacement Glass Flow Cell
For Use With (Equipment) Conductivity Probes (013016MD, 013016A and 013016D)
Type Replacement Flow Cell
NIST Traceable Yes
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.