missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes

Catalog No. 13642255
Change view
Click to view available options
Type:
Epoxy Refillable Triode
Epoxy Triode Low Maintenance
For Use With (Equipment):
Orion A+ Series Meters
Orion Star Series Meters
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
13642255 Epoxy Triode Low Maintenance Orion Star Series Meters
13640 Epoxy Triode Low Maintenance Orion A+ Series Meters
13642256 Epoxy Refillable Triode Orion Star Series Meters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13642255 Supplier Thermo Scientific™ Supplier No. 9179BNMD

Accurately measure ORP and temperature together in just one electrode, with a built-in temperature sensor.

Thermo Scientific™ Orion™ Triode 3-in-1 pH/Automatic Temperature Compensation Electrodes come with a built-in ATC and are engineered with an epoxy body for ruggedness. The 9179BN and 9179BNMD are low-maintenance gel-filled electrodes, best suited for educational institutes where minimal maintenance is required, while the 9180BNMD is a refillable electrode best suited for general purpose samples.
 

Orion™ Single Junction Electrodes provide a reliable option for routine ORP measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required:

Electrode fill solution for 9180BNMD

  • 900011 ORP filling solution for samples with total ionic strength > 0.2 M, 5 x 60 mL bottles
  • 900001 ORP filling solution for samples with total ionic strength < 0.2 M, 5 x 60 mL bottles

Electrode storage solution

  • 910001 4 M KCl storage solution, 475 mL bottle

Standards

  • 967901 ORP standard, 475 mL bottle
  • 967961 ORP standard, 5 x 60 mL bottles

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with 9810BNMD

  • 900011 ORP filling solution for samples with total ionic strength > 0.2 M, 60 mL 1 bottle

Warranty period

  • 9179BN and 9179BNMD: 6 months from date of purchase
  • 9180BNMD: 12 months from date of purchase

Specifications

Applications Water testing,wastewater, disinfection, aquaculture, process water.
Connector Types BNC (Waterproof)
Description Epoxy Triode Low Maintenance
Material Epoxy
Certifications/Compliance ASTM D1498, Standard Methods 2580, USGS Ch 6.5
For Use With (Equipment) Orion Star Series Meters
Temperature (Metric) Operating 0°C to 80°C
Type Epoxy Triode Low Maintenance