Learn More
Thermo Scientific™ Orion™ Pro Star Series Meter Accessories
Select Thermo Scientific™ Orion™ Pro Star series meter accessories to support your Orion Pro Star PH211, EC212, DO213, PI214, PC215, PD216, PI217 and MP218 bench meters.
$81.59 - $815.85
Specifications
| For Use With (Equipment) | Orion Pro Star series bench electrochemistry meters |
|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|
01107437
|
Thermo Scientific™
PSTARARM |
Electrode Stand/Holder |
Each for $407.93
|
|
|||||
|
01107438
|
Thermo Scientific™
PSTARPCR |
Rugged Protective Display Cover |
Each for $815.85
|
|
|||||
|
01107439
|
Thermo Scientific™
PSTARPCS |
Standard Protective Display Cover |
Each for $95.11
|
|
|||||
|
01107440
|
Thermo Scientific™
PSTARPWR |
Power Supply |
Each for $117.74
|
|
|||||
|
01107441
|
Thermo Scientific™
PSTARSHH |
Holder Head |
Each for $117.74
|
|
|||||
|
01107442
|
Thermo Scientific™
PSTARSWB |
Weighted Base |
Each for $140.35
|
|
|||||
|
01107443
|
Thermo Scientific™
PSTARUBS |
USB Barcode Scanner |
Each for $526.28
|
|
|||||
|
01107444
|
Thermo Scientific™
PSTARUFD |
USB Flash Drive |
Each for $81.59
|
|
|||||
|
01107445
|
Thermo Scientific™
PSTARUKB |
USB Keyboard |
Each for $140.35
|
|
|||||
|
01107446
|
Thermo Scientific™
PSTARUMS |
USB Mouse |
Each for $149.46
|
|
|||||
|
01107447
|
Thermo Scientific™
PSTARUSB |
USB Computer Cable |
Each for $81.59
|
|
|||||
Description
Select Orion Pro Star series meter accessories to support your Orion Pro Star PH211, EC212, DO213, PI214, PC215, PD216, PI217 and MP218 bench meters. Add an optional accessory to input sample ID with the USB barcode scanner, USB keyboard or USB mouse. Replace included meter accessories such as the universal power adapter, USB flash drive, USB computer cable and electrode stand with base. Select a standard or rugged protective display cover to add additional protection for your meter in challenging lab conditions.
Select Orion Pro Star series meter accessories to support your Orion Pro Star bench meters. All listed accessories are compatible with the following meter models:
- Orion Pro Star PH211 pH/ORP bench meter
- Orion Pro Star EC212 conductivity bench meter
- Orion Pro Star DO213 dissolved oxygen bench meter
- Orion Pro Star PI214 pH/ion bench meter
- Orion Pro Star PC215 pH/ion and conductivity bench meter
- Orion Pro Star PD216 pH/ion and DO bench meter
- Orion Pro Star PI217 dual channel pH/ion bench meter
- Orion Pro Star MP218 pH/ion, conductivity and DO bench meter
Orion Pro Star Series Meter Optional Accessories
- PSTAR-UBS Orion Pro Star series USB barcode scanner is tested to be compatible with Orion Pro Star meters and allows sample ID values to be easily entered into the meter from barcoded samples.
- PSTAR-UFD Orion Pro Star series USB flash drive is tested to be compatible with Orion Pro Star meters and allows data to be transferred from the meter to a computer, methods to be transferred between meters and software updates to be transferred from the web to the meter.
- PSTAR-UKB Orion Pro Star series USB keyboard is tested to be compatible with Orion Pro Star meters and allows various numeric and alphanumeric values including sample ID values to be easily entered into the meter.
- PSTAR-UMS Orion Pro Star series USB mouse is tested to be compatible with Orion Pro Star meters and allows the meter to be operated with a remote device, removing the need to touch the display to operate the meter, which can be helpful in environments where sample carryover may damage the meter.
- PSTAR-PCR Orion Pro Star series rugged protective display cover is designed to add additional protect to the meter display during operation, which can be helpful in environments where the display may become dirty or scratched in the lab.
Orion Pro Star Series Meter Replacement Accessories
- PSTAR-ARM Orion Pro Star series electrode stand with base allows you to replace your complete electrode stand as needed.
- PSTAR-SHH Orion Pro Star series holder head allows you to replace just the holder head component of the electrode stand as needed.
- PSTAR-SWB Orion Pro Star series weighted base allows you to replace just the base of the electrode stand as needed.
- PSTAR-PCS Orion Pro Star series standard protective display cover allows you to replace you display cover as needed.
- PSTAR-PWR Orion Pro Star series universal power supply allows you to replace your power supply as needed.
- PSTAR-USB Orion Pro Star series USB computer cable allows you to replace your meter-to-computer cable as needed.
Additional Accessories Compatible with Orion Pro Star Series Meters
- 096019-WA Orion stirrer probe for bench meters
- STARA-106 Orion compact ink printer with meter-to-printer cable, ink, paper and 100-240V 50/60 Hz power supply
Specifications
| Orion Pro Star series bench electrochemistry meters |