Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Supplier: Thermo Scientific™ AC4P79
Description
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
- Equivalent to other widely used and recognized chemistries
- Compatible with Orion™ AQUAfast™ Colorimeters
Chemical Identifiers
| Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter | |
| Powder Pack | |
| 100 tests | |
| AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Specifications
| Iron (total) | |
| 100 Iron TPTZ Powder Packs | |
| 100 tests | |
| AQUAfast 24 mm round vials (Cat. No. AC2V24) |
| Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter | |
| Powder Pack | |
| 0.02 to 1.8 mg/L Fe | |
| TPTZ |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.