Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Catalog No. 13642210
Change view
Click to view available options
Type:
Ammonia
Chlorine (free)
Chlorine (total)
Copper (free)
Iron (ferro)
Manganese
Nitrite
Phosphate (ortho)
Sulfate
Range:
0.01 to 0.3 mg/L N
0.01 to 0.8 mg/L N
0.02 to 2 mg/L Cl2
0.02 to 3 mg/L Fe
0.05 to 5 mg/L Cu
0.06 to 2.5 mg/L PO4
0.1 to 18 mg/L Mn
5 to 100 mg/L SO4
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range
13642210 Ammonia 0.01 to 0.8 mg/L N
13642221 Copper (free) 0.05 to 5 mg/L Cu
13642215 Nitrite 0.01 to 0.3 mg/L N
13642217 Manganese 0.1 to 18 mg/L Mn
13642220 Chlorine (free) 0.02 to 2 mg/L Cl2
13642211 Chlorine (total) 0.02 to 2 mg/L Cl2
13642219 Iron (ferro) 0.02 to 3 mg/L Fe
13642212 Sulfate 5 to 100 mg/L SO4
13642214 Phosphate (ortho) 0.06 to 2.5 mg/L PO4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13642210 Supplier Thermo Scientific™ Supplier No. AC4P12
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters
Specifications
Type Ammonia
EPA Approved Methods Wastewater: Standard Methods 4500-NH3 F-1997
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
Includes Set of 2: 200 Ammonia Salicylate Powder Packs, 200 Ammonia Cyanurate Powder Packs
Packaging Powder Pack
Quantity 100 tests
Range 0.01 to 0.8 mg/L N
Recommended Accessories AQUAfast 24 mm round vials (Cat. No. AC2V24)
Test Method Salicylate
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.