Learn More
Thermo Scientific™ Orion™ Star A Series Portable Meter pH Electrode Holder
Catalog No. 13645600
Change view
Click to view available options
pH Electrode Holder
Catalog No.
13645600 pH Electrode Holder
1 options
Catalog No. 13645600 Supplier Thermo Scientific™ Supplier No. STARA ESPH
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter pH Electrode Holder with Star A series portable meter armor and sleeve to protect your pH electrode in the field
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter pH Electrode Holder with the Star A series portable meter armor (Cat. No. STARA-AR) and storage sleeve (Cat. No. 810017) to protect, transport and store your sensors in the field. This pH electrode holder can be attached to either side of the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeve.
Specifications
Description Orion Star A series pH electrode holder for portable meter protective armor
Type Meter Accessories
Recommended Accessories pH electrode storage sleeve (Cat. No. 810017)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.