missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Electrode Filling Solution

Catalog No. 13642402
Change view
Click to view available options
Filling Solution Type:
Ag/AgCl Fill solution
AgCl Fill Solution
KCl Fill Solution
pH Electrode Fill
Reference Inner Fill
Reference Outer Fill
ROSS Electrode Fill
Sodium micro electrode fill
Quantity:
5/Cs.
1 x 60 mL
5 x 60 mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filling Solution Type Quantity
13642402 pH Electrode Fill 5 x 60 mL
13641755 ROSS Electrode Fill 5 x 60 mL
13641899 Ag/AgCl Fill solution 5 x 60 mL
13641901 Reference Inner Fill 5 x 60 mL
13641897 Reference Outer Fill 5 x 60 mL
13641685 Sodium micro electrode fill 1 x 60 mL
13641686 AgCl Fill Solution 5 x 60 mL
13641845 pH Electrode Fill 5/Cs.
13200312 KCl Fill Solution 5 x 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13642402 Supplier Thermo Scientific™ Supplier No. ORI510011

Use this pH electrode filling solution with pH electrodes and reference electrodes.

Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes. Packaged as five 2 oz. (5 x 60mL) bottles.

  • Only use ROSS™ filling solution (810007) with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
  • When the single junction reference electrode (900100) or double junction reference electrode (900200) are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide

 

Specifications

Description Filling Solution for No Cal pH Electrodes
For Use With (Equipment) No Cal pH Electrodes
Type pH Electrode Filling Solution
Filling Solution Type pH Electrode Fill
Packaging Bottle
Quantity 5 x 60 mL

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.