Learn More
Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.
Supplier:  Thermo Scientific™ 911125
 View more versions of this product
Catalog No. 13641277
Description
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.
Cleaning Solutions:
  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
Specifications
Chemical Identifiers
pH Electrode
Solutions restore proper electrode performance and prolong the useful life of the electrode
Specifications
25 Pack of pH Electrode Rinse Solution Pouches
Rinse Solution
Solutions restore proper electrode performance and prolong the useful life of the electrode
25 pk.
pH Electrode
Rinse
Pouch
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.