missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ PerpHecT™ ROSS™ Combination pH Micro Electrode PROMO

Measure micro volume samples with this refillable, glass double junction electrode.

Supplier:  Thermo Scientific™ 8220BNWPPM

Catalog No. 13-642-800PM

Only null left
Explore available promotions

Description

Description

Includes:

  • 1m cable

Requires:

  • pH Meter
  • Electrode
  • Beakers - plastic or glass
  • Magnetic stirrer
  • Required solutions
  • Two pH buffers: pH 7 and pH 4 /10
  • ROSS internal filling solution: 3M KCl; Orion Cat. No. 810007

Compatible with:

  • Orion™ PerpHecT™ meters
Specifications

Specifications

Combination pHmicro Electrode
Glass Double
0°C to 100°C
Waterproof BNC connector; Glass double junction; Refillable
To ensure a quick response and free-flowing liquid junction the sensing element and reference junction must not be allowed to dry out. Short-term Storage (up to one week): Soak electrode in ROSS pH Electrode Storage Solution, Orion 810001 or use pH Electrode Storage Solution, Orion 910001. Tominimize the chance of breakage the 8220 BNWP should be attached to an electrode stand and suspended in a beaker that contains pH electrode storage solution. The electrode should not touch the sides or bottom of the beaker. Long-term Storage (more than one week): The reference chamber should be filled and the filling hole securely covered. Cover the sensing element and reference Junction with the protective cap/sleeve containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
1 m
BNC (Waterproof)
Glass
Less than 0.002 pH units/day
4.5 mm
155 mm
3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.