Learn More
Thermo Scientific™ Orion™ Ion Selective Electrode Accessories
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
$315.01 - $1,735.60
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity & Availability  
13642260
View Documents Promotion Details
Thermo Scientific™
9300BNWP
ISE Half Cell Electrodes Handle
Each for $1,735.60
Only null left
13641771
View Documents Promotion Details
Thermo Scientific™
948201
Ion Selective Electrode Polishing Strips
Pack of 24 Each for $315.01
Only null left
Description
Description
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Electrodes handles are used with plastic membrane combination or half-cell electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes.
Videos
SDS
Documents
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.