Learn More
Thermo Scientific™ Orion™ Iodide Ion Selective Electrodes
Get fast, stable and reliable iodide ions readings in aqueous solutions.
$2,149.41 - $2,544.06
Specifications
For Use With (Equipment) ISE Meter
View More Specs
Products 2
Catalog Number Mfr. No. Connector Types Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types Price Quantity & Availability  
1329991
Accessories View Documents Promotion Details
Thermo Scientific™
9453BN
BNC
Each for $2,149.41
Only null left
13645160
Accessories View Documents Promotion Details
Thermo Scientific™
9653BNWP
BNC (Waterproof)
Each for $2,544.06
Only null left
Description
Description
Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes with built-in Thermo Scientific™ Sure-Flow™ reference. Combination design decreases the amount of required sample and reduces waste.
Features:
  • 9653BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
  • Iodide Half-Cell Electrode 9453BN is to be used with the Orion 900200 sure flow double Junction electrode.
Solutions required:
  • Standard
    • 945306 0.1 M iodide standard, 475 mL bottle
  • ISA
    • 940011 ISA for iodide measurements, 475 mL bottle
  • Electrode fill solution
    • 900063 Optimum Results D electrode filling solution, 5 x 60 mL bottles
    • 900002 Inner fill solution for double junction ref electrode, 5 x 60 mL bottles
    • 900003 Outer fill solution for double junction ref electrode, 5 x 60 mL bottles
Items included with:
  • 9653BNWP
    • 900063 Optimum Results D electrode filling solution, 60 mL 1 bottle
    • 948201 Polishing strip. Qty 2.
    9453BN
    • 948201 Polishing strip. Qty 1.
Warranty period
  • 12 months from date of purchase
Specifications
Specifications
ISE Meter