Learn More
Thermo Scientific™ Orion™ Cupric Ion Selective Electrodes
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically.
Specifications
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Combination ISE
View More Specs
Products 2
Catalog Number Mfr. No. Connector Types Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types Price Quantity & Availability  
1329988
Accessories View Documents Promotion Details
Thermo Scientific™
9429BN
BNC N/A
Only null left
13645157
Accessories View Documents Promotion Details
Thermo Scientific™
9629BNWP
BNC (Waterproof) N/A
Only null left
Description
Description
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically with the Thermo Scientific™ Orion™ Cupric Electrodes.
Features:
  • 9629BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
  • Cupric half cell electrodes 9429BN and 9429SC are to be used with the Orion 900200 sure flow double junction electrode.
  • 9429SC requires a separate electrode cable with a screw cap connector.
Solutions required:
  • Standards
    • 942906 0.1 M cupric standard, 475 mL bottle
  • ISA
    • 940011 ISA for cupric measurements, 475 mL bottle
  • Electrode fill solution
    • 900002 Inner chamber filling solution for the double junction reference electrode, 5 x 60 mL bottles
    • 900003 Outer chamber filling solution for the double junction reference electrode, 5 x 60 mL bottles
    • 900063 Optimum Results D electrode filling solution, 5 x 60 mL bottles
Items included with:
  • 9429BN & 9429SC
    • 948201 Polishing strip. Qty 2.
  • 9629BNWP
    • 900063 Optimum Results D electrode filling solution, 60ml 1 bottle
    • 948201 Polishing strip. Qty 1.
Warranty period
  • 12 months from date of purchase
Specifications
Specifications
Sure-Flow Junction
ISE Meter
Combination ISE