Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Cadmium Combination Ion Selective Electrode
Catalog No. 13645159
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Catalog No. Connector Types
13645159 BNC (Waterproof)
1 options
Catalog No. 13645159 Supplier Thermo Scientific™ Supplier No. 9648BNWP
Measure cadmium ions in aqueous solutions quickly, simply, accurately and economically.
Measure cadmium ions in aqueous solutions quickly, simply and economically with the Thermo Scientific™ Orion™ Cadmium Electrodes.
Features:
  • 9648BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
Solutions required:
  • ISA
    • 940011 ISA for cadmium measurements,475 mL bottle
  • Electrode fill solution
    • 900061 Optimum Results A electrode filling solution, 5 x 60 mL bottles
Items included with:
  • 900061 Optimum Results A electrode filling solution. 60 mL 1 bottle
  • 948201 Polishing strip. Qty 1.
Warranty period
  • 12 months from date of purchase
Specifications
Connector Types BNC (Waterproof)
Description Model 9448BN; Orion Cadmium Combination
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Combination ISE