missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis Spectrophotometers

Catalog No. 15200946
Change view
Click to view available options
Type:
7100 Vis Spectrophotometer
8100 UV-Vis Spectrophotometer
Plug Type:
NA, EU, UK Power cord
Spectral Bandwidth:
2 nm
5 nm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Plug Type Spectral Bandwidth
15200946 8100 UV-Vis Spectrophotometer NA, EU, UK Power cord 2 nm
15200945 7100 Vis Spectrophotometer NA, EU, UK Power cord 5 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15200946 Supplier Thermo Scientific™ Supplier No. AQ8100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Test Tube Holder for vials and test tube from 12 to 25 mm OD, 10 mm square single-cell holder, AquaMate Water Analysis Methods Software, Snap-on Printer, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362)

Optimized for your water analysis needs AquaMate 7100 Vis and AquaMate 8100 UV-Vis Spectrophotometers provide fast and reliable results

Thermo Scientific™ Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogrammed methods, and are designed to meet the needs of water and wastewater analysis laboratories. AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation.

Pre-programmed Methods:

  • More than 260 preprogrammed methods offer easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Preprogrammed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries

Versatility:

  • Glove friendly 7-inch touch screen controller
  • Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates vials and test tube from 12 to 25 mm OD

Verification:

  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
  • Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
  • Built-in test methods for stray light, noise and resolution are available to validate instrument performance further

Compliance

CE / TUV

Warranty

1 Year

Specifications

Baseline Flatness ±0.002 A
Certifications/Compliance CE / TUV
Compartments Accessible from top, front or side Able to accommodate cells up to 100 mm cells Available accessory for test tubes up to 25 mm diameter and 150 mm height Removable, washable sample compartment liner with magnetic placement and hold-down
Connections Single USB-A
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Depth (Metric) 35.5 cm
Detector Type Dual Silicon Photodiodes
Display 7 in. High Definition Color Touchscreen, Fixed, 800 x 1280 pixels
Dimensions (L x W x H) 35.5 x 38.5 x 19.5 cm
Spectral Bandwidth 2 nm
Type 8100 UV-Vis Spectrophotometer
Wavelength Range 190 to 1100 nm
Drift <0.0005 A/hr. (At 500 nm after 1 hour warmup)
Electrical Requirements External AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz
For Use With (Application) Water and Waste Water Analysis
Height (Metric) 19.5 cm
Includes Test Tube Holder for vials and test tube from 12 to 25 mm OD, 10 mm square single-cell holder, AquaMate Water Analysis Methods Software, Snap-on Printer, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362).
Interface Glove friendly touch-screen
Keypad Glove- friendly touchscreen
Lamp Life >5 years typical
Length (Metric) 35.5 cm
Noise (RMS at 500 nm 60 consecutive measurements) ≤0.00020A at 0A at 260 and 500 nm ≤0.00030A at 1A at 260 and 500 nm ≤0.00040A at 2A at 260 and 500 nm
Optical Design Double Beam
Photometric Accuracy ±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A
Photometric Display -3A to +5A
Photometric Range -2A to +3.5A
Photometric Repeatability ±0.001A at 1A measured at 1.0A at 546 nm
Plug Type NA, EU, UK Power cord
Printer Included Snap-on printer included
Scan Speed Slow, medium and fast - up to 1600 nm/min.
Stray Light < 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Warranty 1 Year
Wavelength Accuracy ±0.5 nm
Wavelength Data Interval 0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability < ±0.2 nm
Weight (Metric) 7.5 kg
Width (Metric) 38.5 cm
Show More Show Less