Learn More
Thermo Scientific™ Orion™ AquaMate™ Power Cords
Catalog No. 13650708
Change view
Click to view available options
Voltage:
110 V
Plug Type:
US Plug
Catalog No. Voltage Plug Type
13650708 110 V US Plug
1 options
Catalog No. 13650708 Supplier Thermo Scientific™ Supplier No. AQ110V
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use Thermo Scientific™ Orion™ AquaMate Power Cords with AquaMate 8000 UV-Vis and AquaMate 7000 Vis spectrophotometers
Use Thermo Scientific™ Orion™ AquaMate Power Cords with AquaMate 8000 UV-Vis and AquaMate 7000 Vis spectrophotometers. Select power cords with a US / Japan plug, Euro plug, UK / Singapore plug or China / Australia / New Zealand plug.
AquaMate Power Cords include the AquaMate 110V US, Japan Plug Power Cord (Cat. No. AQ110V) and AquaMate 220V Euro Plug Power Cord (Cat. No. AQ220V). The AquaMate 240V UK, Singapore Plug Power Cord (Cat. No. AQ240V) and AquaMate China, Australia, New Zealand Plug Power Cord (Cat. No. AQCHNV). Purchase separately.
Specifications
Product Line AquaMate
Type Power Cord
Voltage 110 V
Plug Type US Plug
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.