Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Catalog No. 13642170
Change view
Click to view available options
Type:
Ammonia
Chlorine
Iron
Nitrate
Nitrite
Sulfate
Zinc
Range:
0.08 to 1 mg/L N
0.2 to 1.5 mg/L N
0.2 to 3 mg/L N
0.2 to 6 mg/L Fe
0.3 to 3 mg/L Zn
0.4 to 5 mg/L Cl2
10 to 100 mg/L SO4
Method:
Bariumsulfate-Turbidity
Cadmium Reduction
Diazotization (Azo)
DPD
Phenanthroline
Salicylate
Zincon
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range Method
13642170 Nitrate 0.2 to 1.5 mg/L N Cadmium Reduction
361005101 Ammonia 0.2 to 3 mg/L N Salicylate
13642173 Nitrite 0.08 to 1 mg/L N Diazotization (Azo)
13642182 Zinc 0.3 to 3 mg/L Zn Zincon
13642161 Chlorine 0.4 to 5 mg/L Cl2 DPD
13642167 Iron 0.2 to 6 mg/L Fe Phenanthroline
13642180 Sulfate 10 to 100 mg/L SO4 Bariumsulfate-Turbidity
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13642170 Supplier Thermo Scientific™ Supplier No. AC4004
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifications
Type Nitrate
Test Reagent Formats Ampoule
For Use With (Equipment) Orion AQUAfast AQ4000 Colorimeter
Description AQUAfast nitrate LR Auto-ID ampoules, 30 pack
Includes 30 Auto-ID Ampoules
Method Cadmium Reduction
For Use With (Application) Colorimetric water and wastewater testing equipment
Recommended Accessories AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
Range 0.2 to 1.5 mg/L N
EPA Approved Methods Drinking Water: ASTM D3867-90 (B) / Standard Methods 4500-NO3- E; Wastewater: ASTM D3867-04 (B) / Standard Methods 4500-NO3- E
Quantity 30 Ampoules
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.