Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Supplier:  Thermo Scientific™ AC4078
 View more versions of this product
Catalog No. 13642167
Only null left
Description
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifications
Specifications
Iron
Orion AQUAfast AQ4000 Colorimeter
30 Auto-ID Ampoules
Colorimetric water and wastewater testing equipment
0.2 to 6 mg/L Fe
30 Ampoules
Ampoule
AQUAfast iron (total & soluble) Auto-ID ampoules, 30 pack
Phenanthroline
AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
Wastewater: Standard Methods 3500-Fe-1997
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.