missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits

Catalog No. 13642689
Change view
Click to view available options
Type:
Alkalinity
Aluminum
Ammonia
Chloride
Chlorine
Copper
Nitrite
Ozone
pH
Phosphate
Silica
Sulfate
Sulfide
Zinc
Range:
0.01 to 0.3 mg/L Al
0.01 to 0.5 mg/L N
0.01 to 6 mg/L Cl2
0.02 to 1 mg/L N
0.02 to 1 mg/L Zn
0.04 to 0.5 mg/L S
0.05 to 0.5 mg/L O3
0.05 to 4 mg/L PO4
0.05 to 4 mg/L SiO2
0.05 to 5 mg/L Cu
0.5 to 25 mg/L Cl-
1 to 80 mg/L PO4
5 to 100 mg/L SO4
5 to 200 mg/L CaCO3
6.5 to 8.4 pH
Quantity:
100 DPD No 1 Tablets
100 DPD No 4 Tablets
100 Tablets
100 Tests
50 Tests
50 Tests Each (free and total)
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range Quantity
13642689 Chlorine 0.01 to 6 mg/L Cl2 50 Tests Each (free and total)
13642000 Alkalinity 5 to 200 mg/L CaCO3 100 Tests
13642001 Aluminum 0.01 to 0.3 mg/L Al 50 Tests
13642012 Ammonia 0.02 to 1 mg/L N 50 Tests
13642003 Chloride 0.5 to 25 mg/L Cl- 50 Tests
13642004 Chlorine 0.01 to 6 mg/L Cl2 100 DPD No 1 Tablets
13642005 Chlorine 0.01 to 6 mg/L Cl2 100 DPD No 4 Tablets
13642007 Copper 0.05 to 5 mg/L Cu 50 Tests
13642023 Zinc 0.02 to 1 mg/L Zn 50 Tests
13642014 Nitrite 0.01 to 0.5 mg/L N 100 Tests
13642015 Ozone 0.05 to 0.5 mg/L O3 100 Tests
13642016 pH 6.5 to 8.4 pH 100 Tests
13642017 Phosphate 0.05 to 4 mg/L PO4 50 Tests
13642018 Phosphate 1 to 80 mg/L PO4 50 Tests
13642019 Silica 0.05 to 4 mg/L SiO2 50 Tests
13642020 Silica 0.05 to 4 mg/L SiO2 100 Tablets
13642021 Sulfate 5 to 100 mg/L SO4 100 Tests
13642022 Sulfide 0.04 to 0.5 mg/L S 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 13642689 Supplier Thermo Scientific™ Supplier No. AC2070
Only null left

Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.

Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100

Specifications

Type Chlorine
Test Reagent Formats Tablet
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
Description AQUAfast chlorine (free & total) reagent tablets, 50 tests each
Includes Set of 2: 50 DPD No 1 Tablets, 50 DPD No 3 Tablets
Method DPD
For Use With (Application) Colorimetric water and wastewater testing equipment
Recommended Accessories AQUAfast 24mm round vials (Cat. No. AC2V24)
Range 0.01 to 6 mg/L Cl2
EPA Approved Methods Drinking Water: Standard Methods 4500-Cl G; Wastewater: Standard Methods 4500-Cl G-2000
Quantity 50 Tests Each (free and total)
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.