Learn More
Thermo Scientific™ Orion™ AQUAfast™ Free & Total Chlorine Bulk DPD Powder Reagents
Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials are designed for high volume testing of residual chlorine in water samples.
Supplier:  Thermo Scientific™ AQ250T
 View more versions of this product
Catalog No. 13301156
Only null left
Description
Description
Measure residual chlorine in water samples using Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials with Orion AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. The powder chemistries dissolve quickly in the solution and are easy to mix. Each vial contains 250 counts of DPD reagent and is available for free chlorine or total chlorine analysis in the lab or field.
  • Measure 0.02 to 2.00 mg/L (ppm) free chlorine or total chlorine
  • Bulk powder dispensers are designed for high sample throughput – reduces the cost per test and minimizes waste compared with individually packaged reagents 
  • Equivalent to other widely used and recognized chemistries including Orion AQUAfast AC4P71 free chlorine and Orion AQUAfast AC4P72 total chlorine powder packs
  • Compatible with Orion AQUAfast AQ4000, AQ3700 and AQ3170 colorimeters
  • Compatible with Orion AquaMate AQ8000 and AQ7000 spectrophotometers
Specifications
Chemical Identifiers
0.02 to 2 mg/L Cl2
Powder
Chlorine
DPD
Specifications
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter, Orion AQUAfast AQ3170 Colorimeter
250 count
Powder
DPD
AQ250T Dispenser, 1 vial
0.02 to 2 mg/L Cl2
Chlorine
Product Suggestions
Product Suggestions