missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Single Junction Micro Stem Glass Bodied Combination pH Electrode
Measure small volumes down to 0.5 μL with micro pH electrodes.
Supplier: Thermo Scientific™ 9810BN
Description
Thermo Scientific™ Orion™ Silver Chloride Micro Electrodes provide reliable and accurate measurements of routine samples. Best suited to measure samples as small as 0.5 μL and for use in NMR tubes.
Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required
Electrode fill solution
900011 4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001 pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
electrode storage solution, 475ml, and 12 mm electrode storage bottle
electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
900011 4M KCl with Ag/AgCl filling solution 60mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
| Micro pH Electrode | |
| Ceramic | |
| 0°C to 80°C | |
| 4M KCl with Ag/AgCl, Orion Cat. No. 900011 | |
| Single junction Ag/AgCl | |
| 32°F to 176°F | |
| 3.28 ft. | |
| 1 m |
| BNC | |
| Small Tip Glass | |
| Small tip glass combination micro electrode; Tip: 37 X 1.3mm; Min. sample volume: 0.5μL | |
| For pH measurements in micro volume samples | |
| To ensure a quick response and free-flowing junction, the sensing element and reference junction must not dry out. Always clean the electrode before storing it. Short-term storage (up to one week): Soak the electrode in pH electrode storage solution, Cat. No. 910001. To prevent crystallization of the filling solution, cover the fill hole whenever the electrode is being stored and open the fill hole when calibrating and measuring. Long-term storage (more than one week): Fill the electrode and securely cover the filling hole. Cover the electrode with the protective glass tube containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode. | |
| 4.72 in. | |
| 120 mm |