missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Carbon Dioxide Combination Ion Selective Electrode

Catalog No. 13645156
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13645156 BNC (Waterproof)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13645156 Supplier Thermo Scientific™ Supplier No. 9502BNWP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Measure carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly, accurately and economically.

Measure carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly and accurately with the Thermo Scientific™ Orion™ 9502BNWP Carbon Dioxide Electrode. 

Features:

  • It features a gas-permeable membrane technology for use with loose membranes and O-rings.

Solutions required:

  • Standard
    • 950206 0.1 M NaHCO3 Carbon Dioxide standard, 475 mL
    • 950207 1000 ppm as CaCO3 Carbon Dioxide standard, 475 mL
  • Carbon dioxide buffer solution
    • 950210 Carbon Dioxide Ionic Strength Adjuster (ISA), 475 mL
  • Electrode fill solution
    • 950202 Carbon Dioxide electrode fill solution, 60 mL
  • Storage solution
    • 941706 0.1 M NaCl Chloride standard, 475 mL

Items included with:

  • 950202 Carbon Dioxide electrode fill solution, 60 mL 1 bottle
  • 950204 4 membranes with 1 O-ring

Warranty period:

  • 12 months from date of purchase

Specifications

Connector Types BNC (Waterproof)
Description Carbon Dioxide Electrode
Junction Annular PTFE
Material Epoxy
For Use With (Equipment) ISE Meter with a BNC Connection
Temperature (Metric) Operating 0°C to 50°C
Type Carbon Dioxide Electrode