missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ ROSS™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode
Description
Thermo Scientific™ Orion™ ROSS Flat Surface Electrodes have a uniquely designed, flat sensing bulb that allow for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates.
The 8135BN ROSS Combination Flat Surface pH Electrode measures pH of soft, moist surfaces — both solid and semi-solid —such as agar gel plates, meats, bread dough and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50°C or more.
- Material: Epoxy body
- pH range: 0 to 14
- Temperature range: 0° to 100°C
- Internal reference: ROSS
- Junction: Glass fiber
- Dimensions: 120 × 12mm
- pH precision: 0.01
- Fill solution: Orion 810007
- Cap diameter: 16mm
- Small samples are available in micro sample dish
- Flat surface tip
Includes: One 60 ml bottle of ROSS fill Solution, 810007 and spout cap
Specifications
Specifications
| Description | With BNC connector |
| Connector Types | BNC |
| Junction | Glass Fiber |
| Material | Epoxy Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | Combination Flat Surface pH Electrode, BNC connector |
| Drift | <0.002pH/day |
| Filling Solution Type | 3M KCl |
| For Use With (Equipment) | Standard pH Meter, BNC connector |
| Show More |