missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode

Catalog No. 13641807
Change view
Click to view available options
Connector Types:
BNC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13641807 BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13641807 Supplier Thermo Scientific™ Supplier No. 8135BN
Only null left

Measure flat, soft, moist, semi-solid or small volume samples with epoxy body flat surface pH electrodes.

Thermo Scientific™ Orion™ ROSS Flat Surface Electrodes have a uniquely designed, flat sensing bulb that allow for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates.
 

The 8135BN ROSS Combination Flat Surface pH Electrode measures pH of soft, moist surfaces — both solid and semi-solid —such as agar gel plates, meats, bread dough and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50°C or more.

  • Material: Epoxy body
  • pH range: 0 to 14
  • Temperature range: 0° to 100°C
  • Internal reference: ROSS
  • Junction: Glass fiber
  • Dimensions: 120 × 12mm
  • pH precision: 0.01
  • Fill solution: Orion 810007
  • Cap diameter: 16mm
  • Small samples are available in micro sample dish
  • Flat surface tip

Includes: One 60 ml bottle of ROSS fill Solution, 810007 and spout cap

Specifications

Description With BNC connector
Connector Types BNC
Junction Glass Fiber
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Flat Surface pH Electrode, BNC connector
Drift <0.002pH/day
Filling Solution Type 3M KCl
For Use With (Equipment) Standard pH Meter, BNC connector
Internal Reference ROSS
Precision 0.01
Storage Requirements Short-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Temperature (English) Operating 32°F to 212°F
Length (English) Cable 39 in.
Length (Metric) Cable 1 m
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Diameter (Metric) Cap 16 mm
Show More Show Less