missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS™ Glass Bodied Combination pH Electrode

Catalog No. p-4529992
Change view
Click to view available options
Connector Types:
BNC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13641729 BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13641729 Supplier Thermo Scientific™ Supplier No. 8102BN
Only null left

Capture accurate measurements of a variety of sample types, including solvents, with Thermo Scientific™ Orion™ glass body pH electrodes.

Thermo Scientific™ Orion™ ROSS Refillable Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. They are available in chemically resistant glass bodies and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries.
 

Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
 
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
 
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
 
Warranty period
12 months from date of purchase

Specifications

Connector Types BNC
Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination pH Electrode, BNC connector
Drift <0.002 pH/day
Filling Solution Type 3M KCl
For Use With (Equipment) Standard pH Meter, BNC connector
Includes One 60mL bottle of Orion ROSS filling solution and spout cap
Internal Reference ROSS
Precision 0.01
Storage Requirements Short-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Temperature (English) Operating 32°F to 212°F
Length (English) 4.72 in.
Length (English) Cable 39.37 in.
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Diameter (Metric) Cap 16 mm
Show More Show Less