missing translation for 'onlineSavingsMsg'
Learn More

Orienteering Compass and Kits

Catalog No. S96996
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S96996 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96996 Supplier Customer Channels Group (Ccg) Supplier No. 78530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Compass, red base, lanyard and instructions

Basic, accurate compass to get started with map and compass work.

Compass available individually, as part of an orienteering kit, or as part of a demonstration kit.

Orienteering Compass

  • Graduated in 2° increments and features four scales
  • Black bezel with easy-to-read print
  • Has liquid-filled capsule, declination correction scale, luminous points, and 4X magnifier in base
  • Acrylic base offers scales in inches, centimeters, 1:25,000, and 1:50,000
Teacher's Compass
  • Nonfunctional, oversized 9 × 15 in. (22.8 × 38cm)
  • Clear; can be used on an overhead projector

Specifications

Includes Compass, red base, lanyard and instructions
For Use With (Application) Accurate compass to get started with map and compass work
Increments 2°
Quantity Each
Product Type Orienteering Compass
Material Acrylic