Learn More
Organomation Dry Bath 24 Position N-EVAP Nitrogen Evaporator
Supplier:  Organomation 11250DA
Encompass
The dry bath 24 position N-EVAP is our most adaptable benchtop evaporator. The 24 position N-EVAP offers the most customizable options for your sample preparation needs. The standard sample holder can accommodate samples with an outside diameter of 10-30 mm. However, with the optional 8 position large sample holder, vials with an outside diameter of 30-70 mm can be processed. The 24 position N-EVAP test tube evaporator can also be upgraded to include digital controls for heat and time with the addition of the side control box (Option code -CB).These economical units provide controlled concentration of sample vials through the application of nitrogen gas within a dry heated bath. Which uses aluminum or glass beads for heating. The dual band spring hoist assembly provides a simple and effective means of removing samples from the heated dry bath.
Catalog No. NC0134711
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.