missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Standard, 10,000 ppm C (Ethylene Glycol in 0.2% H2SO4), Ricca Chemical
Supplier: Ricca Chemical Company R18499504C
Specifications
| Organic Carbon Standard | |
| Amber Glass Bottle | |
| Odorless | |
| C8H5KO4 | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
| 204.222 |
| 10,000mg/L C, 10,000ppm C (Ethylene Glycol in 0.2% Sulfuric Acid) | |
| 877-24-7,7664-38-2,7732-18-5 | |
| 10, 000 ± 50ppm | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate | |
| 23676735 |
Chemical Identifiers
| 877-24-7 | |
| 204.222 | |
| 23676735 | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
| C8H5KO4 | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate |