Learn More
Ores of Common Metals
15 large ore samples.
Supplier:  Geoscience Industries SR2251
Numéro de catalogue. S45025
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Recognize metals as they appear naturally
  • Labeled with a descriptive key
  • Represents the most commonly used ores, including aluminum, copper, iron, lead, manganese, and molybdenum
Spécifications
Spécifications
15/Pk.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS