missing translation for 'onlineSavingsMsg'
Learn More

Orange II, Spectrum™ Chemical

$231.96 - $466.14

Chemical Identifiers

CAS 633-96-5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18612090
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $231.96
Only null left
 
18612089
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $466.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

633-96-5

Specifications

633-96-5
Reagent
Orange II
1
Orange/Red
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.