missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orange G, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Color | Orange/Red |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 452.38 |
| Grade | Reagent |
| Chemical Name or Material | Orange G |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.