MW 452.38 g/mol. Azo dye used as a tracking dye for nucleic acids in gel electrophoresis. Used in SDS-capillary electrophoresis as a standard. Also used as a histological stain and as a counterstain for nuclear stains.
The product is subject to the following: Abcam Restricted Use Statement