missing translation for 'onlineSavingsMsg'
Learn More

Orange G, Electrophoresis Grade

Catalog No. AAJ62743A1
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62743A1 1 kg
AAJ6274314 25 g
AAJ6274322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62743A1 Supplier Thermo Scientific Chemicals Supplier No. J62743A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Collagen stain for connective tissue | CAS: 1936-15-8 | C16H10N2Na2O7S2 | 452.36 g/mol

Collagen stain for connective tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1936-15-8
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.36
MDL Number MFCD00012457
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym C.I. 16230; Acid Orange 10
PubChem CID 131850556
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Beilstein 4120705
Color Orange to Red
Solubility Information Soluble in water or methyl cellosolve; Slightly soluble in ethanol
Quantity 1 kg
Physical Form Powder
IUPAC Name disodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Formula Weight 452.38
Grade Electrophoresis
Chemical Name or Material Orange G
EINECSNumber 217-705-6
RTECSNumber QJ6500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only