missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Optochin Disc

Catalog No. OXDD0001T
Change view
Click to view available options
Packaging Type:
1 Cartridge/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
OXDD0001T 1 Cartridge/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDD0001T Supplier Thermo Scientific™ Supplier No. DD0001T
Only null left

Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.

Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.

Ordering Alerts:

5μg. 50 Discs/Cartridge. 1 Cart/Pk.

Specifications

Antimicrobial Agents Optochin (Ethylhydrocuprein Hydrochloride)
Concentration 5 μg
Product Type Optochin Disc
Test Type Microbial Differentiation
Material Paper
Packaging Type 1 Cartridge/Pk.
Quantity 50 Discs/Cartridge
Species Streptococcus pneumoniae