missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ OptiPRO™ SFM AGT™ Medium

Catalog No. A4000164001
Change view
Click to view available options
Quantity:
10 L
100 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4000164001 10 L
A4000164002 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4000164001 Supplier Gibco™ Supplier No. A4000164001
Only null left

OptiPRO™ SFM AGT™ is a serum free culture medium in a easy-to-use granular format for growth of several kidney derived cell lines MDBK, MDCK, PK-15, and VERO.

Gibco OptiPRO SFM AGT Medium is a flexible-format medium that is equivalent to OptiPRO SFM with the addition of Advanced Granulation Technology for simplified preparation. OptiPRO SFM is a serum-free, animal-origin-free culture medium designed for growth of several kidney-derived cell lines, including MDBK, MDCK, PK-15, and VERO, for virus or recombinant protein production. In addition, OptiPRO SFM has been used to successfully grow other cell lines, such as Hela, BHK-21, and COS-7.

Features of OptiPRO SFM AGT Medium include:

  • Serum-free, animal-origin-free, low-protein formulation
  • Easy adaptation for many popular cell lines
  • Scalability in flasks, cell factories, and bioreactors with AGT

Serum-free, animal-origin-free, low-protein formulation

OptiPRO SFM is formulated with a very low protein concentration (< 10 μg/mL), for easier purification of recombinant proteins.

Easy adaptation for many popular cell lines

OptiPRO SFM was designed for growth of kidney-derived cell lines such as MDCK, MDBK, VERO, and PK-15. But it has also been used successfully to grow other cell lines, such as Hela, BHK-21, and COS-7, allowing the use of a single medium for multiple cell lines. OptiPRO SFM is formulated without L-glutamine for greater stability and extended shelf life. Simply add L-glutamine or Glutamax supplement as needed.

cGMP manufacturing and quality system

OptiPRO SFM is manufactured in a cGMP-compliant facility. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Scalability with Advanced Granulation Technology

Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration, a just-add-water protocol, and low dust generation, creating less pressure build-up during sterile filtration.

Specifications

Cell Line BHK-21, Vero, MDCK, MDBK, PK-15, COS-7, HeLa
Cell Type Mammalian
Classification Animal Origin, Low Protein, Serum-free, cGMP
Concentration 1X
Form Powder
Product Type Media
Serum Level Serum-free
Sterility Non-sterile
Sterilization Method Non-sterile
With Additives Phenol Red, Sodium Pyruvate, Sodium Bicarbonate, Thymidine
Without Additives No Glutamine
Quantity 10 L
Formulation Proprietary
Culture Type Adherent Cell Culture
Expression System Mammalian
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.