missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ OptiPRO™ SFM
GREENER_CHOICE

Catalog No. p-7076129
Encompass
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12309050 100 mL
12309019 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12309050 Supplier Gibco™ Supplier No. 12309050
Only null left

OptiPRO™ SFM is a serum-free, animal origin-free culture medium designed for growth of several kidney-derived cell lines including MDBK, MDCK, PK-15 and VERO for virus or recombinant protein production.

OptiPRO™ SFM is a serum-free, animal origin-free culture medium designed for growth of several kidney-derived cell lines including MDBK, MDCK, PK-15 and VERO for virus or recombinant protein production. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7. Gibco™ OptiPRO™ SFM features:

  • Serum-free, animal origin-free, low protein formulation
  • Direct adaptation for many popular cell lines

Serum-free, animal origin-free, low protein formulation

Gibco™ OptiPRO™ SFM is formulated with a very low protein concentration (< 10 μg/mL), allowing for easier purification of your recombinant protein.

Direct adaptation for many popular cell lines

Gibco™ OptiPRO™ SFM has been designed for growth of kidney-derived cell lines such as MDCK, MDBK, VERO and PK-15. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7, allowing you to choose a single medium for multiple cell lines.

cGMP Manufacturing and Quality System

Gibco™ OptiPRO™ SFM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Cell Line HeLa, MDCK, BHK-21, PK-15, COS-7, MDBK, VERO
Classification Animal Origin-free, Low Protein, Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium
Serum Level Serum-free
With Additives Phenol Red, Sodium Pyruvate, Sodium Bicarbonate, Thymidine, Hypoxanthine
Without Additives No Glutamine
Product Line Gibco
Quantity 100 mL
Shipping Condition Room Temperature
Endotoxin Level Low
Format Bottle(s)
Included Antibiotics No antibiotics
Content And Storage Storage conditions: 2 – 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.