missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Set of Four Heratrays (1/2 width)

Catalog No. 11688019
Change view
Click to view available options
Material:
Copper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11688019 Copper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11688019 Supplier Thermo Scientific™ Supplier No. 50065810

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Material: Copper; For Use With: Heracell 240i

Specifications

For Use With (Equipment) Heracell 240i
Lighting LED
Material Copper
Type Tray
Width (English) 0.5 in.
Width (Metric) 1.27 cm