missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSA OptimAir™ TL PAPR Standard Kit
Description
- Available with facepiece or hood
- Motor blower electronics allow for optimum airflow
- Compact and ergonomic battery pack available in standard or extended-life
- Automatic flow rates
- Crushproof breathing tube and sealing gaskets on cartridges
- Audible and visual alarms for low flow indication and low battery voltage
- Suitable applications include chemical, hazardous materials, asbestos, mold and lead paint abatement, and pharmaceutical
Certifications
NIOSH approved.
Specifications
Specifications
| Battery Type | Nickel Hydride |
| Certifications (NIOSH) | NIOSH |
| Product Type | PAPR Assembly with Hood |
| Facepiece/Headtop Model | Hood |
| For Use With (Application) | Chemical, hazardous materials, asbestos, mold and lead paint abatement, and pharmaceutical |
| For Use With (Equipment) | TL powered air purifying respirator |
| Includes | Motor blower, smart charger, standard battery, hose and decon belt |
| Intrinsic Safety (IS) Rating | No |
| Model | 10081116 |
| Weight (English) | 5.76 lb. |
| Show More |