missing translation for 'onlineSavingsMsg'
Learn More

Carson™ Optigami Build Your Own Cardboard Microscope Kit

Catalog No. S98214
Encompass
Change view
Click to view available options
For Use With (Equipment):
Specimen Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S98214 Specimen Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD: Small parts. Not for children under 3 years.
Catalog No. S98214 Supplier Carson™ Supplier No. EPDOG100
CHOKING HAZARD: Small parts. Not for children under 3 years.

Experience the fun of assembling your very own microscope with the Optigami Build-Your-Own Cardboard Microscope Kit. The Optigami will provide hours of entertainment! It’s a great way to get the whole family involved and nurture a child’s love of science.

  • Use as a stand-alone compound microscope or attach the smartphone platform to make a digital microscope to help capture photos or videos of your exciting finds
  • Have fun viewing the included prepared slides
  • With a powerful 50x to 400x magnification, discover things the naked eye can't see
  • Powered by 2 AA batteries (sold separately)
  • Fits all phones

Specifications

Type Cardboard Microscope
Viscosity 100 to 120 mPas
Autoclavable Not Autoclavable
Color Multi-colored
Depth (English) 6.6 in.
Depth (Metric) 16.8 cm
For Use With (Application) Viewing Intricate Details
For Use With (Equipment) Specimen Slides
Height (English) 10 in.
Height (Metric) 25.4 cm
Includes 6 Cardboard Assembly Sheets, 1 Optical Magnification Scope (50x to 400x Magnification), 6 Thick Rubber Bands, 1 LED Module, 1 Focus Control Tube, 2 Blank Microscope Slides with Coverslips, 13 Prepared Slides, and 1 Eye Dropper
Length (English) 5.6 in.
Length (Metric) 14.2 cm
Material Cardboard
Required Accessories 2 AA Batteries (Sold Separately)
Sterility Non-sterile
Weight (English) 0.59 lb.
Weight (Metric) 267.6 g
Width (English) 6.6 in.
Width (Metric) 16.8 cm
Show More Show Less