missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Optifit Tips, Refills
GREENER_CHOICE

Catalog No. 14559462
Change view
Click to view available options
Color Code:
Blue
Gray
Green
Sterility:
Non-sterile
Sterile
Volume (Metric):
0.1 to 10 μL
10 to 1000 μL
100 to 5000 μL
Packaging:
Bulk in Carton
Refill Pack
Refill Tower
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code Sterility Volume (Metric) Packaging
14559462 Gray Non-sterile 0.1 to 10 μL Refill Tower
14559480 Blue Sterile 10 to 1000 μL Refill Pack
14559479 Blue Non-sterile 10 to 1000 μL Refill Pack
14559498 Green Non-sterile 100 to 5000 μL Bulk in Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14559462 Supplier Sartorius Supplier No. 790012
Only null left

Ensure perfect fitting and leak-tight sealing on Sartorius pipettes. Sartorius Optifit Tips, Refills promote maximum pipetting accuracy and reproducibility. Tips are manufactured in an ISO 8 certified cleanroom environment and are available in sterile and non-sterile versions in a variety of volumes.

Ergonomically designed for maximum precision and accuracy
  • Perfect tip fitting and sealing to the pipetter
  • Enables ergonomic and light tip attachment and ejection
  • Precision made tips provide best possible accuracy and precision
Compatible with Optiload feature in Sartorius pipetters and more:
  • Picus, eLINE™, mLINE™ and Proline™ Plus
  • Color-coded trays and tips to easily match with compatible Sartorius pipetters
  • Universal design is compatible with most other brands of pipetters
Designed to the highest standards
  • Made from high-quality virgin polypropylene
  • Refill Packs are DNase, RNase and endotoxin-free certified
  • Available as e-beam pre-sterilized or non-sterilized
  • Improved tip identification and traceability by informative rack labels
    • Indicates tip volume, product number, lot number and purity level
  • Packaging and tips are 100% recyclable or can be incinerated
  • Available in a range of packaging formats:
    • Refill Packs - purity certified and ecological packages
    • Refill Towers - space-saving and ecological
    • Bulk Cartons - economical bulk boxes
  • Air-tight plastic wrapping around the rack secures purity during transport and storage
  • Reusable and fully autoclavable at 121°C for 20 minutes

Certifications

CE certified. Manufactured according to ISO 9001 and ISO 13485.

Specifications

Volume (Metric) 0.1 to 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Transparent
Autoclavable Autoclavable
Length (English) 1.24 in.
Length (Metric) 3.15cm
Certifications/Compliance CE, ISO 9001, ISO 13485
Sterilization Method Autoclave at 121°C
Format Rack
Tip Style Standard
Packaging Refill Tower
Product Type Non-filtered Tips
Color Code Gray
Disposable Single-use
For Use With (Equipment) Sartorius Picus, Sartorius eLINE, Sartorius mLINE, Sartorius Proline Plus
Graduated No
Product Line Optifit
Material Polypropylene
Show More Show Less