missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Optifit Tips Easybulk Pack

Manufactured under ISO Class 8 Cleanroom conditions. The orderly arrangement of tips allows the packs to be approximately 40% smaller than conventional bulk tip cartons, resulting in more space on your workbench and reduced packaging waste by nearly 50%.

Supplier:  Sartorius LHEB791204

 View more versions of this product

Catalog No. 14558736


Only null left
Explore available promotions

Description

Description

  • Perfect fit and seal with Sartorius pipettes to ensure the highest possible accuracy and precision
  • Ergonomic and light tip attachment and ejection enabled by the Optiload feature in Sartorius Picus™ NxT, Picus™, Tacta™, mLINE™ and Proline™ Plus pipettes
  • Compatibility with most other pipette brands
  • Compliance with ISO 9001, ISO 14001, and ISO 13485 quality standards
  • Specifications

    Specifications

    1200 μL
    Non-sterile
    White / Clear
    Optifit tips
    50-1200 μl
    ISO 9001, ISO 14001 and ISO 13485
    Universal
    480
    Single-use
    No
    Polypropylene
    Non-filtered
    Clear
    Not Autoclavable
    50-1200 μl
    71.5 mm
    Bulk
    Bulk carton
    Non-filtered Tips
    Universal Fit
    Optifit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions