missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Optifit Extended Plus Tip
GREENER_CHOICE

Catalog No. 12133944
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
12133944 Non-sterile
12133945 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12133944 Supplier Sartorius Supplier No. LHX791000

The Optifit Extended Plus tips are designed to improve pipetting reach, making them ideal for use with deep and narrow labware.

  • The Optifit Extended Plus tips are designed to improve pipetting reach, making them ideal for use with deep and narrow labware
  • The tip design allows for more efficient retrieval of samples, down to the last drop
  • The longer length of these tips helps prevent contamination by ensuring that the pipette does not come into contact with the walls of labware or containers during use
  • The tips are manufactured in an ISO 8 certified cleanroom environment and are certified to be free of DNase, RNase, endotoxin and human DNA
  • Sartorius pipettes and tips are designed to seamless fit together for the most reliable and reproducible results

Specifications

Volume (Metric) 1 mL
Filtered Non-filtered
Sterility Non-sterile
Color Transparent
Autoclavable Autoclavable
Format Rack
Tip Style Ergonomic
Product Type Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less