Learn More
United Scientific™ Optics Set
Catalog No. S66968
Change view
Click to view available options
Material:
Acrylic
Catalog No. Material
S66968 Acrylic
1 options
Catalog No. S66968 Supplier United Scientific™ Supplier No. OPSET1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore the world of light reflection and defraction with the Optics Set.
  • Basic optics set can be used by itself or with the current optics experiments.
  • Two acrylic lenses (one double convex and one double concave).
  • Three mirrors (one convex, one concave and one plane).
  • Comes in a wood storage box.
Specifications
Quantity 1 Ea.
Material Acrylic
Includes Two Acrylic lenses, one each of double convex and double concave, three mirrors, one each of convex, concave and plane
For Use With (Application) Explore the world of light reflection and defraction.
Product Type Optics Set
Shape Various
Dimensions (L x W x H) 12.7 x 8.9 x 3.8 cm (5 x 3.5 x 1.5 in.)